Learn More
Cytiva PhastGel™ Buffer Strips
Pre-prepared 3% agarose blocks containing buffer, for use in conjunction with PhastGel gradient and homogeneous gel media to generate a multiphasic buffer system during electrophoresis
£81.10 - £81.65
Specifications
For Use With (Equipment) | PhastSystem Automated Electrophoresis Unit |
---|---|
For Use With (Application) | Gel Electrophoresis |
Product Code | Brand | Quantity | For Use With (Application) | Product Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | For Use With (Application) | Product Type | Price | Quantity & Availability | ||||||
10269294
|
Cytiva
18-1668-01 |
2 Pack | Gel Electrophoresis | Buffer strip holder | N/A | N/A | N/A | |||||
11584915
|
Cytiva
17-0517-01 |
20 Strips | Gel Electrophoresis | PhastGel Buffer strips-native | N/A | N/A | N/A | |||||
11798448
|
Cytiva
17-0516-01 |
20 Strips | Gel Electrophoresis | PhastGel Buffer strips-SDS | N/A | N/A | N/A | |||||
Description
- Ready made buffer strips eliminate the need to prepare electrophoresis buffers
- Supplied as 10X pairs - for both cathode and anode
- Choice of Native or Denaturing (SDS) buffer systems
Specifications
PhastSystem Automated Electrophoresis Unit | |
Gel Electrophoresis |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Cytiva PhastGel™ Buffer Strips