missing translation for 'onlineSavingsMsg'
Learn More

Phencyclone, 97%, Thermo Scientific Chemicals

Product Code. 11332665
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5660-91-3
Molecular Formula C29H18O
Molecular Weight (g/mol) 382.46
MDL Number MFCD00051451
InChI Key MNSDGJFEKUKHGO-UHFFFAOYSA-N
Synonym phencyclone, 1,3-diphenyl-2h-cyclopenta l phenanthren-2-one, 1,3-diphenylcyclopenta l phenanthren-2-one, 1,3-diphenylcyclopenta 1,2-l phenanthren-2-one
PubChem CID 97140
ChEBI CHEBI:48304
IUPAC Name 1,3-diphenylcyclopenta[l]phenanthren-2-one
SMILES O=C1C(=C2C(=C1C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C21)C1=CC=CC=C1

Specifications

Melting Point 245°C to 249°C
Quantity 1 g
Beilstein 2061817
Formula Weight 382.46
Percent Purity 97%
Chemical Name or Material Phencyclone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'