missing translation for 'onlineSavingsMsg'
Learn More

Phenethyleisothiocyanate, 97%, Thermo Scientific™

Product Code. 10216004
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 2257-09-2
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.24
MDL Number MFCD00004821
InChI Key IZJDOKYDEWTZSO-UHFFFAOYSA-N
Synonym 2-phenylethyl isothiocyanate, phenethyl isothiocyanate, phenylethyl isothiocyanate, 2-isothiocyanatoethyl benzene, phenethyl mustard oil, phenylaethylsenfoel, benzene, 2-isothiocyanatoethyl, peitc, phenylethyl mustard oil, 2-phenylethylisothiocyanate
PubChem CID 16741
ChEBI CHEBI:351346
IUPAC Name 2-isothiocyanatoethylbenzene
SMILES C1=CC=C(C=C1)CCN=C=S

Specifications

CAS Min % 97.0
Density 1.1000g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula C6H5CH2CH2NCS
Quantity 1 g
Merck Index 15, 7340
Formula Weight 163.24
Percent Purity 97%
Chemical Name or Material Phenethyl isothiocyanate
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'