missing translation for 'onlineSavingsMsg'
Learn More

Phenol/Chloroform/Isoamyl Alcohol (25:24:1 Mixture, pH 5.2±0.3, Liq.), Fisher BioReagents

£62.74 - £172.83

Specifications

Chemical Name or Material Phenol/Chloroform/Isoamyl Alcohol
Boiling Point 96°C
Molecular Formula CHCl3
MDL Number MFCD00000826
Merck Index 15, 7354
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10081403
View Documents Promotion Details
Fisher BioReagents
BP1753I-100
100 mL Coated Amber Glass/Poison Pack This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10295103
View Documents Promotion Details
Fisher BioReagents
BP1753I-400
400 mL Coated Amber Glass/Poison Pack This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This mixture is used for extracting protein from nucleic acid preparations.
Specifications

Specifications

Phenol/Chloroform/Isoamyl Alcohol
CHCl3
15, 7354
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane
≥99.0%
Coated Amber Glass/Poison Pack
Liquid
96°C
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
ClC(Cl)Cl
119.37
Pass Test
5.2
Videos
Safety and Handling

Safety and Handling

missing translation for 'dotInformation' : DOT Class 6.1, : Poison

Recommended Storage : 4°C

SDS
Documents

Documents

Product Certifications