Learn More
Thermo Scientific Chemicals Phenol:Chloroform:Isoamyl alcohol 25:24:1, Ready-to-Use saturated aq. soln., pH 6.7, with alkaline buffer
Useful for purifying RNA
£100.50 - £241.20
Chemical Identifiers
CAS | 136112-00-0 |
---|---|
Molecular Formula | C12H19Cl3O2 |
Molecular Weight (g/mol) | 301.632 |
MDL Number | MFCD00133763 |
InChI Key | ZYWFEOZQIUMEGL-UHFFFAOYSA-N |
Synonym | phenol-chloroform-isoamyl alcohol mixture, chloroform; isoamyl alcohol; phenol, phenol chloroform isoamyl alcohol, phenol-chloroform-isoamyl alcohol mixture, bioultra, for molecular biology, 125:24:1, phenol-chloroform-isoamyl alcohol mixture, bioultra, for molecular biology, 25:24:1, phenol-chloroform-isoamyl alcohol mixture, vetec tm reagent grade, 25:24:1, phenol-chloroform-isoamyl alcohol mixture, bioultra, for molecular biology, 49.5:49.5:1, phenol:chloroform:isoamyl alcohol 25:24:1, ready-to-use saturated aqueous solution, ph 5.2, with alkaline buffer, phenol:chloroform:isoamyl alcohol 25:24:1, ready-to-use saturated aqueous solution, ph 6.7, with alkaline buffer |
PubChem CID | 66587205 |
IUPAC Name | chloroform;3-methylbutan-1-ol;phenol |
SMILES | CC(C)CCO.C1=CC=C(C=C1)O.C(Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15455785
|
Thermo Scientific Chemicals
J60331.AE |
100 mL |
£100.50
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15465785
|
Thermo Scientific Chemicals
J60331.AN |
400 mL |
£241.20
400mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phenol:Chloroform:Isoamyl alcohol 25:24:1 is used in the extraction of DNA for the generation of DNA minisatellite fingerprints. It is also used for purifying RNA.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
136112-00-0 | |
301.632 | |
ZYWFEOZQIUMEGL-UHFFFAOYSA-N | |
66587205 | |
CC(C)CCO.C1=CC=C(C=C1)O.C(Cl)(Cl)Cl |
C12H19Cl3O2 | |
MFCD00133763 | |
phenol-chloroform-isoamyl alcohol mixture, chloroform; isoamyl alcohol; phenol, phenol chloroform isoamyl alcohol, phenol-chloroform-isoamyl alcohol mixture, bioultra, for molecular biology, 125:24:1, phenol-chloroform-isoamyl alcohol mixture, bioultra, for molecular biology, 25:24:1, phenol-chloroform-isoamyl alcohol mixture, vetec tm reagent grade, 25:24:1, phenol-chloroform-isoamyl alcohol mixture, bioultra, for molecular biology, 49.5:49.5:1, phenol:chloroform:isoamyl alcohol 25:24:1, ready-to-use saturated aqueous solution, ph 5.2, with alkaline buffer, phenol:chloroform:isoamyl alcohol 25:24:1, ready-to-use saturated aqueous solution, ph 6.7, with alkaline buffer | |
chloroform;3-methylbutan-1-ol;phenol |
Specifications
Colorless | |
C12H19Cl3O2 | |
UN2922 | |
ZYWFEOZQIUMEGL-UHFFFAOYSA-N | |
chloroform;3-methylbutan-1-ol;phenol | |
66587205 | |
Liquid |
136112-00-0 | |
MFCD00133763 | |
phenol-chloroform-isoamyl alcohol mixture, chloroform; isoamyl alcohol; phenol, phenol chloroform isoamyl alcohol, phenol-chloroform-isoamyl alcohol mixture, bioultra, for molecular biology, 125:24:1, phenol-chloroform-isoamyl alcohol mixture, bioultra, for molecular biology, 25:24:1, phenol-chloroform-isoamyl alcohol mixture, vetec tm reagent grade, 25:24:1, phenol-chloroform-isoamyl alcohol mixture, bioultra, for molecular biology, 49.5:49.5:1, phenol:chloroform:isoamyl alcohol 25:24:1, ready-to-use saturated aqueous solution, ph 5.2, with alkaline buffer, phenol:chloroform:isoamyl alcohol 25:24:1, ready-to-use saturated aqueous solution, ph 6.7, with alkaline buffer | |
CC(C)CCO.C1=CC=C(C=C1)O.C(Cl)(Cl)Cl | |
301.632 | |
6.7 | |
Phenol:Chloroform:Isoamyl alcohol 25:24:1, Ready-to-Use saturated aq. soln., with alkaline buffer |
Safety and Handling
GHS H Statement
H331-H314-H341-H351-H373-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
Recommended Storage : 2° to 8°C
RUO – Research Use Only