Learn More
Phenol:Chloroform Saturated Solution, pH 5.2, MP Biomedicals
This is a ready-to-use saturated solution of phenol and chloroform suitable for RNA purifications. 1:1 solution, pH 5.2
Brand: MP Biomedicals 04802515
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 2821 Chem class : 6.1 Pack group : II
Specifications
Phenol:Chloroform Saturated Solution | |
400 mL |