missing translation for 'onlineSavingsMsg'
Learn More

Phenol, Fused Crystal, White, Ultrapure Bioreagent, J.T. Baker™

Product Code. 15578224
Click to view available options
Quantity:
100 g
500 g
Packaging:
Steel Can
This item is not returnable. View return policy
This item is not returnable. View return policy

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Extraction of Nucleic Acids and Molecular Biology Applications

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Color White
Flash Point 79.00°C
Packaging Steel Can
Quantity 100 g
Specific Gravity 1L = 1.06kg
Formula Weight 94.11
Grade Ultrapure Bioreagent
Physical Form Fused Crystals
Chemical Name or Material Phenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'