missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Phenol Red Product Code.: 11480307

Thermo Scientific Chemicals Phenol Red

Product Code. 11480307
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11480307

Brand: Thermo Scientific Chemicals B21710.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phenol red is used as a non absorbed marker in gravimetric method. It is a pH indicator used in cell biology laboratories and in home swimming pool test kits. It is used to check the kidney function based on the excreted phenol red by colorimetric method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenol red is used as a non absorbed marker in gravimetric method. It is a pH indicator used in cell biology laboratories and in home swimming pool test kits. It is used to check the kidney function based on the excreted phenol red by colorimetric method.

Solubility
Soluble in water, ethanol and methanol. Insoluble in chloroform.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-74-8
Molecular Formula C19H14O5S
Molecular Weight (g/mol) 354.38
MDL Number MFCD00003552
InChI Key BELBBZDIHDAJOR-UHFFFAOYSA-N
Synonym phenol red, phenolsulfonphthalein, fenolipuna, sulfonphthal, sulphental, sulphonthal, phenolsulphonphthalein, psp indicator, phenolsulfonephthalein, unii-i6g9y0j1oj
PubChem CID 4766
ChEBI CHEBI:31991
SMILES OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Boiling Point >300°C
Odor Odorless
Quantity 50 g
Beilstein 326470
Merck Index 14,7247
Solubility Information Soluble in water,ethanol and methanol. Insoluble in chloroform.
IUPAC Name 4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
Formula Weight 354.38
Chemical Name or Material Phenol Red

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253532 Certificate TypeCertificate of Analysis Date24/12/2024 Catalog NumberB21710.30,B21710.18,B21710.09
Lot Number10251114 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberB21710.30,B21710.18,B21710.09
Lot Number10248977 Certificate TypeCertificate of Analysis Date14/12/2023 Catalog NumberB21710.30,B21710.18,B21710.09
Lot NumberX18I001 Certificate TypeCertificate of Analysis Date31/10/2022 Catalog NumberB21710.30,B21710.18,B21710.09
Lot NumberS29I018 Certificate TypeCertificate of Analysis Date11/07/2022 Catalog NumberB21710.30,B21710.18,B21710.09
compliance-icons
Product Identifier
  • Phenol red
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Phenolsulfonphthalein

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Phenol Red >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.