missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenol Red

Product Code. 10299473
Click to view available options
Quantity:
5 g
10 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-74-8
Molecular Formula C19H14O5S
Molecular Weight (g/mol) 354.38
MDL Number MFCD00003552
InChI Key BELBBZDIHDAJOR-UHFFFAOYSA-N
Synonym Phenolsulfonephthalein
PubChem CID 4766
ChEBI CHEBI:31991
SMILES OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Melting Point >300.0°C
Clarity passes test
Beilstein 19, 91
Merck Index 15, 7360
Solubility Information Solubility in water: 0.77g/L. Other solubilities: soluble in ethanol
Infrared Spectrum Authentic
IUPAC Name 3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Formula Weight 354.38
Grade ACS Reagent
Packaging Glass bottle
Visual Transition Interval from pH 6.8 (yellow) to pH 8.2 (red)
Color Colorless to Yellow
Quantity 100 g
Physical Form Liquid After Melting
Chemical Name or Material Phenol Red, Pure
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'