missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Phenol Red, Indicator Product Code.: 10665202

Thermo Scientific Chemicals Phenol Red, Indicator

Product Code. 10665202
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10665202

Brand: Thermo Scientific Chemicals 151430250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-74-8
Molecular Formula C19H14O5S
Molecular Weight (g/mol) 354.38
MDL Number MFCD00003552
InChI Key BELBBZDIHDAJOR-UHFFFAOYSA-N
Synonym Phenolsulfonephthalein
PubChem CID 4766
ChEBI CHEBI:31991
SMILES OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Melting Point >300.0°C
Beilstein 19, 91
Merck Index 15, 7360
Solubility Information Solubility in water: 0.77g/L. Other solubilities: soluble in ethanol
Infrared Spectrum Authentic
IUPAC Name 3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Formula Weight 354.38
Grade Pure
Packaging Glass bottle
Specific Extinction 1700 (558nm, in buffer) (E (1% min./1cm))
Visual Transition Interval from pH 6.8 (yellow) to pH 8.2 (red)
Color Beige/Brown to Gray
Quantity 25 g
Physical Form Crystalline Powder, Crystals, Flakes and/or Chunks
Chemical Name or Material Phenol Red
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0441565 Certificate TypeCertificate of Analysis Date23/02/2024 Catalog Number151431000,151430250,151430050
Lot NumberA0459055 Certificate TypeCertificate of Analysis Date09/01/2024 Catalog Number151431000,151430250,151430050
Lot NumberA0395168 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number151431000,151430250,151430050
Lot NumberA0413301 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number151431000,151430250,151430050
Lot NumberA0432122 Certificate TypeCertificate of Analysis Date16/07/2021 Catalog Number151431000,151430250,151430050
compliance-icons
Product Identifier
  • Phenol Red
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Phenol Red, Indicator >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.