missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenol Red, pure, indicator

Product Code. p-180951 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10791745

Brand: Thermo Scientific Chemicals 151430050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 143-74-8 | C19H14O5S | 354.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-74-8
Molecular Formula C19H14O5S
Molecular Weight (g/mol) 354.38
MDL Number MFCD00003552
InChI Key BELBBZDIHDAJOR-UHFFFAOYSA-N
Synonym Phenolsulfonephthalein
PubChem CID 4766
ChEBI CHEBI:31991
SMILES OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Melting Point >300.0°C
Beilstein 19, 91
Merck Index 15, 7360
Solubility Information Solubility in water: 0.77g/L. Other solubilities: soluble in ethanol
Infrared Spectrum Authentic
IUPAC Name 4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
Formula Weight 354.38
Grade Pure
Packaging Glass bottle
Specific Extinction 1700 (558nm, in buffer) (E (1% min./1cm))
Visual Transition Interval from pH 6.8 (yellow) to pH 8.2 (red)
Color Beige/Brown to Gray
Quantity 5 g
Physical Form Crystalline Powder, Crystals, Flakes and/or Chunks
Chemical Name or Material Phenol Red
Show More Show Less
compliance-icons
Product Identifier
  • Phenol Red
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Phenolsulfonphthalein

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.