missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol Red Solution, Fisher Chemical™
Click to view available options
Quantity:
100 mL
Specifications
Color | Red |
pH | 2.1 to 2.2 |
Quantity | 100 mL |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Phenol red solution. European Pharmacopoeia Ref: 1063601 (RP 146)
- Danger
- Flammable liquid Category 3
- Serious eye damage/eye irritation Category 2
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'