Learn More
Phenol, Saturated (pH 6.6/7.9, Liq.), Fisher BioReagents™
£130.00 - £169.00
Chemical Identifiers
CAS | 108-95-2, 6381-92-6, 77-86-1 |
---|---|
Molecular Formula | C6H6O |
Molecular Weight (g/mol) | 94.11 |
MDL Number | MFCD00002143 |
InChI Key | ISWSIDIOOBJBQZ-UHFFFAOYSA-N |
Synonym | carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol |
PubChem CID | 996 |
ChEBI | CHEBI:15882 |
IUPAC Name | phenol |
SMILES | OC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10264673
|
Fisher BioReagents
BP1750I-100 |
100 mL | Coated Amber Glass/Poison Pack |
£130.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10001173
|
Fisher BioReagents
BP1750I-400 |
400 mL | Coated Amber Glass/Poison Pack |
£169.00
400mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Phenol, Saturated (pH 6.6/7.9, Liq.)
Phenol saturated with Tris buffer is used in DNA extraction procedures.Chemical Identifiers
108-95-2, 6381-92-6, 77-86-1 | |
94.11 | |
ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
996 | |
phenol |
C6H6O | |
MFCD00002143 | |
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol | |
CHEBI:15882 | |
OC1=CC=CC=C1 |
Specifications
108-95-2, 6381-92-6, 77-86-1 | |
C6H6O | |
2821 | |
ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
phenol | |
996 | |
≥99.0% | |
Liquid | |
DNase- and RNase-Free |
≥99.0 | |
MFCD00002143 | |
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol | |
OC1=CC=CC=C1 | |
94.11 | |
CHEBI:15882 | |
Pass Test | |
Phenol, Saturated |
Safety and Handling
DANGER!
Emergency Overview
Combustible liquid. Causes burns by all exposure routes. Possible risks of irreversible effects. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.
NFPA
Health:3
Flammability:2
Instability:0
missing translation for 'dotInformation' : DOT Class 6.1, : Poison
Recommended Storage : 4°C
- Phenol saturated pH 6.6
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- Germ cell mutagenicity Category 2
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H373-May cause damage to organs.
- H341-Suspected of causing genetic defects.
- H311-Toxic in contact with skin.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Phenol, Tris (hydroxymethyl) aminomethane, Ethylenediaminetetraacetic acid, disodium salt dihydrate