missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenol/TRIS saturated sol., for molecular biology, stabilized, DNAse, RNAse and Protease free

Product Code. 10431212 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 10431212

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phenol/TRIS , CAS # 96210-45-6, is a molecular biology solvent system that can be used for the isolation of DNA. | CAS: 108-95-2 | C6H6O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Phenol/Tris is a solvent system consisting of phenol and Tris(Hydroxymethyl)aminomethane
  • Prepared as ready-to-use, no titration necessary

Applications

  • Commonly used in DNA isolation. Tris(Hydroxymethyl)aminomethane acts as a proton acceptor to raise pH
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenol/TRIS saturated sol.
Color Yellow to Orange
Name Note Stabilized, DNase, RNase and Protease Free
CAS 108-95-2, 77-86-1, 7732-18-5
Molecular Formula C6H6O
MDL Number MFCD01632719
Composition Phenol (108-95-2); Tris (hydroxymethyl) aminomethane (77-86-1)
Density 1.07
Merck Index 15, 7354
Formula Weight 94.11
Grade Molecular Biology
pH 7.8 to 8.2 (25°C)
Quantity 500 mL
Physical Form Solution
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenol/TRIS saturated sol.
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.