missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenolphthalein, 98.5%

£35.18 - £402.00

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
PubChem CID 4764
ChEBI CHEBI:34914
IUPAC Name 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10406980
SDS View Documents
Thermo Scientific Chemicals
147711000
100 g Plastic bottle
£35.18
100g
In Stock
to see stock.
Add to basket
 
10626732
SDS View Documents
Thermo Scientific Chemicals
147715000
500 g Plastic bottle
£106.63
500g
In Stock
to see stock.
Add to basket
 
10195343
SDS View Documents
Thermo Scientific Chemicals
147710025
2.5 kg Plastic bottle
£402.00
2.50kg
Estimated Shipment: 31-03-2025
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

77-09-8
318.33
KJFMBFZCATUALV-UHFFFAOYSA-N
CHEBI:34914
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
C20H14O4
MFCD00005913
4764
3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one

Specifications

258.0°C to 263.0°C
98.5%
MFCD00005913
15, 7356
KJFMBFZCATUALV-UHFFFAOYSA-N
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
318.33
CHEBI:34914
0.5% max. (105°C)
Pure
White
Phenolphthalein
77-09-8
C20H14O4
18, 143
Solubility in water: <0.1%. Other solubilities: 1g/12mL alcohol, 1g/100mL ether, soluble in alkalies, soluble in chloroform
Authentic
3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
4764
318.32
98.5%
1.299
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

missing translation for 'einecsNumber' : 201-004-7

missing translation for 'rtecsNumber' : SM8380000

missing translation for 'tsca' : TSCA

Product Identifier
  • Phenolphthalein

Signal Word
  • Danger

Hazard Category
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2

Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H361f-Suspected of damaging fertility.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.