missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Phenolphthalein, ACS Product Code.: 11328977

Thermo Scientific Chemicals Phenolphthalein, ACS

Product Code. 11328977
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11328977

Brand: Thermo Scientific Chemicals 038705.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Indicator

Phenolphthalein is widely used as an indicator in acid-base titrations, water treatment applications. Due to its pH sensitivity, It is used to the check extend of carbonation in concrete, soap and in toys as diapering work. It is used in Kastle-Meyer test, which identifies the substances containing blood.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
PubChem CID 4764
ChEBI CHEBI:34914
IUPAC Name 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Melting Point 258°C to 262°C
Beilstein 284423
Merck Index 14,7243
Solubility Information Soluble in alcohol; Slightly soluble in ether; Slightly soluble in dimethyl sulfoxide and insoluble in benzene or hexane.
Formula Weight 318.33
Grade ACS
Density 1.299 g/mL
Quantity 100 g
Physical Form Powder
Chemical Name or Material Phenolphthalein, For analysis ACS

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM11L520 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number038705.22,038705.36
Lot NumberZ28K520 Certificate TypeCertificate of Analysis Date06/01/2025 Catalog Number038705.22,038705.36
Lot NumberU01K516 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog Number038705.22,038705.36
Lot NumberX25I515 Certificate TypeCertificate of Analysis Date04/10/2023 Catalog Number038705.22,038705.36,038706.AE
Lot NumberR22J527 Certificate TypeCertificate of Analysis Date13/07/2023 Catalog Number038705.22,038705.36
compliance-icons
Product Identifier
  • Phenolphthalein
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H361f-Suspected of damaging fertility.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : phenolphthalein

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Phenolphthalein, ACS >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.