missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenolphthalein disulfate, tripotassium salt trihydrate, 98%, pure

Product Code. p-181028 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 10403171

Brand: Thermo Scientific Chemicals 417220050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62625-16-5 | C20H13K3O11S2 | 610.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62625-16-5
Molecular Formula C20H13K3O11S2
Molecular Weight (g/mol) 610.73
MDL Number MFCD00150194
InChI Key PLHSVCWOEHYSHO-UHFFFAOYSA-K
PubChem CID 129893851
SMILES [K+].[K+].[K+].OC(C1=CC=C(OS([O-])(=O)=O)C=C1)(C1=CC=C(OS([O-])(=O)=O)C=C1)C1=CC=CC=C1C([O-])=O

Specifications

Melting Point 230.0°C
Assay Percent Range 98%
Solubility Information Solubility in water: slightly soluble.
Infrared Spectrum Authentic
IUPAC Name tripotassium 2-{hydroxybis[4-(sulfonatooxy)phenyl]methyl}benzoate
Formula Weight 664.78
Percent Purity 98%
Grade Pure
Packaging Glass bottle
Color White
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Phenolphthalein disulfate, tripotassium salt trihydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.