missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Phenolphthalein, For analysis ACS Product Code.: 10289840

Thermo Scientific Chemicals Phenolphthalein, For analysis ACS

Product Code. 10289840
500 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10289840

Brand: Thermo Scientific Chemicals 417185000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
PubChem CID 4764
ChEBI CHEBI:34914
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Melting Point 258.0°C to 263.0°C
Clarity Passes Test
Beilstein 18, 143
Merck Index 15, 7356
Solubility Information Solubility in water: <0.1%. Other solubilities: 1g/12mL alcohol, 1g/100mL ether, soluble in alkalies, soluble in chloroform
Infrared Spectrum Authentic
IUPAC Name 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
Formula Weight 318.32
Grade ACS Reagent
Packaging Plastic bottle
Vapor Pressure Vapor Pressure: Negligible
Visual Transition Interval from pH 8.0 (colorless) to pH 10 (red)
Density 1.299g/mL
Color White
Quantity 500 g
Physical Form Crystalline Powder
Chemical Name or Material Phenolphthalein
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464551 Certificate TypeCertificate of Analysis Date12/07/2024 Catalog Number417181000,417180025,417180026,417185000
Lot NumberA0456213 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number417181000,417180025,417180026,417185000
Lot NumberA0449971 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number417181000,417180026,417185000
Lot NumberA0396332 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number417181000,417180025,417180026,417185000
Lot NumberA0394481 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number417181000,417180025,417180026,417185000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Phenolphthalein, For analysis ACS >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.