missing translation for 'onlineSavingsMsg'
Learn More
Phenoxazine, 97%, Thermo Scientific Chemicals Product Code.: 10336870

Phenoxazine, 97%, Thermo Scientific Chemicals

Product Code. 10336870
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10336870

Brand: Thermo Scientific Acros 130220010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-67-1
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD00005014
InChI Key TZMSYXZUNZXBOL-UHFFFAOYSA-N
Synonym phenoxazine, phenazoxine, unii-c2zwt499sg, c2zwt499sg, 5,4-oxazine, phenotazine, rarechem fh 2c 0s92, phenoxazine 1g, 9ci
PubChem CID 67278
IUPAC Name 10H-phenoxazine
SMILES N1C2=CC=CC=C2OC2=CC=CC=C12

Specifications

Melting Point 154.0°C to 159.0°C
Color Gray to Gray-Green
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 27, 62
Merck Index 15, 7365
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in abs.methanol, ethanol, ether,, chloroform and benzene, sparingly soluble in petroleum ether
Formula Weight 183.21
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material Phenoxazine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0435364 Certificate TypeCertificate of Analysis Date15/04/2024 Catalog Number130220050,130220250,130220010
Lot NumberA0405860 Certificate TypeCertificate of Analysis Date22/02/2024 Catalog Number130220050,130220250,130220010
Product Identifier
  • Phenoxazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phenoxazine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.