missing translation for 'onlineSavingsMsg'
Learn More

Phenoxyacetyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11474804
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Acylation of D- and DL-WV with phenoxyacetyl chloride, followed by cleavage of the t-butyl ester,afforded the penultimate penicilloic acids. Phenoxyacetyl chloride was used in the synthesis of series of macrocyclic bis-β-lactams, 5-phenyl-6-epiphenoxymethylpenicillin benzyl ester, N-protected guanosine derivatives, useful in RNA synthesis, phenyloxyketene, for cycloaddition to imines leading to β-lactams. Treatment of iminophosphoranes with phenoxyacetyl chloride afforded respectively the trans- and cis-3-(acylamino)-p-lactams. By the oxidative addition of stannous fluoride to allyliodide, with 1, 2-O-isopropylidene-D-glyceraldehyde and phenoxyacetyl chloride results in the predominant formation of erythro homoallylester, which is in turn converted into 2-deoxy-D-ribose. The 4-disubstituted P-lactams (4a-c and 6a-b) were prepared through the reaction of N,N-diphenylhydrazones and N-methyl-N-phenylhydrazones of ketones with phenoxyacetyl chloride/Et3N in dichloromethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acylation of D- and DL-WV with phenoxyacetyl chloride, followed by cleavage of the t-butyl ester,afforded the penultimate penicilloic acids. Phenoxyacetyl chloride was used in the synthesis of series of macrocyclic bis-β-lactams, 5-phenyl-6-epiphenoxymethylpenicillin benzyl ester, N-protected guanosine derivatives, useful in RNA synthesis, phenyloxyketene, for cycloaddition to imines leading to β-lactams. Treatment of iminophosphoranes with phenoxyacetyl chloride afforded respectively the trans- and cis-3-(acylamino)-p-lactams. By the oxidative addition of stannous fluoride to allyliodide, with 1, 2-O-isopropylidene-D-glyceraldehyde and phenoxyacetyl chloride results in the predominant formation of erythro homoallylester, which is in turn converted into 2-deoxy-D-ribose. The 4-disubstituted P-lactams (4a-c and 6a-b) were prepared through the reaction of N,N-diphenylhydrazones and N-methyl-N-phenylhydrazones of ketones with phenoxyacetyl chloride/Et3N in dichloromethane.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 701-99-5
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00000726
InChI Key PKUPAJQAJXVUEK-UHFFFAOYSA-N
Synonym phenoxyacetyl chloride, acetyl chloride, phenoxy, phenyloxyacetyl chloride, phenoxyacetylchloride, phenoxyacetic acid chloride, acetyl chloride, 2-phenoxy, pac-cl, phenoxylacetylchloride, 2-phenoxyacetylchloride, phenoxy acetyl chloride
PubChem CID 69703
IUPAC Name 2-phenoxyacetyl chloride
SMILES ClC(=O)COC1=CC=CC=C1

Specifications

Density 1.235
Boiling Point 225°C to 226°C
Flash Point 108°C (226°F)
Odor Irritating
Refractive Index 1.534
Quantity 1000 g
UN Number UN3265
Beilstein 607585
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 170.6
Percent Purity 98%
Chemical Name or Material Phenoxyacetyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'