missing translation for 'onlineSavingsMsg'
Learn More

Phenyl acetate, 97%, Thermo Scientific Chemicals

Product Code. 10438110
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-79-2
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00008699
InChI Key IPBVNPXQWQGGJP-UHFFFAOYSA-N
Synonym acetic acid phenyl ester, phenol acetate, acetylphenol, acetic acid, phenyl ester, acetyl phenol, acetoxybenzene, acetates, acetic acid,phenyl ester, fenylester kyseliny octove, fenylester kyseliny octove czech
PubChem CID 31229
ChEBI CHEBI:8082
IUPAC Name phenyl acetate
SMILES CC(=O)OC1=CC=CC=C1

Specifications

Color Colorless to Brown
Density 1.0700g/mL
Boiling Point 196.0°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3CO2C6H5
Refractive Index 1.5010 to 1.5030
Quantity 5 g
Beilstein 06,152
Fieser 01,829
Merck Index 15,7379
Specific Gravity 1.07
Solubility Information Solubility in water: ca 4g/L (25°C). Other solubilities: miscible with alcohol,chloroform,ether,soluble in glacial acetic acid
Formula Weight 136.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Phenyl acetate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Phenyl acetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'