Learn More
Phenyl benzoate, 99%, Thermo Scientific Chemicals
£17.19 - £149.75
Chemical Identifiers
CAS | 93-99-2 |
---|---|
Molecular Formula | C13H10O2 |
Molecular Weight (g/mol) | 198.22 |
MDL Number | MFCD00003072 |
InChI Key | FCJSHPDYVMKCHI-UHFFFAOYSA-N |
Synonym | benzoic acid phenyl ester, diphenylcarboxylate, benzoic acid, phenyl ester, benzoic acid phenyl, phenylbenzoate, unii-b8a3wvz590, phenol, benzoate, dsstox_cid_28185, dsstox_rid_82744, dsstox_gsid_48210 |
PubChem CID | 7169 |
ChEBI | CHEBI:86919 |
IUPAC Name | phenyl benzoate |
SMILES | C1=CC=C(C=C1)C(=O)OC2=CC=CC=C2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10592371
|
Thermo Scientific Acros
162060050 |
5 g | Glass bottle |
£17.19
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10296110
|
Thermo Scientific Acros
162061000 |
100 g | Plastic bottle |
£35.68
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10695212
|
Thermo Scientific Acros
162065000 |
500 g | Plastic bottle |
£149.75
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
93-99-2 | |
198.22 | |
FCJSHPDYVMKCHI-UHFFFAOYSA-N | |
7169 | |
phenyl benzoate |
C13H10O2 | |
MFCD00003072 | |
benzoic acid phenyl ester, diphenylcarboxylate, benzoic acid, phenyl ester, benzoic acid phenyl, phenylbenzoate, unii-b8a3wvz590, phenol, benzoate, dsstox_cid_28185, dsstox_rid_82744, dsstox_gsid_48210 | |
CHEBI:86919 | |
C1=CC=C(C=C1)C(=O)OC2=CC=CC=C2 |
Specifications
1mg KOH/g max. | |
68.0°C to 70.0°C | |
298.0°C to 299.0°C | |
98.5% min. (GC) | |
C6H5CO2C6H5 | |
09, 116 | |
benzoic acid phenyl ester, diphenylcarboxylate, benzoic acid, phenyl ester, benzoic acid phenyl, phenylbenzoate, unii-b8a3wvz590, phenol, benzoate, dsstox_cid_28185, dsstox_rid_82744, dsstox_gsid_48210 | |
FCJSHPDYVMKCHI-UHFFFAOYSA-N | |
phenyl benzoate | |
7169 | |
198.22 | |
Fine Crystalline Powder |
93-99-2 | |
White | |
Authentic | |
C13H10O2 | |
MFCD00003072 | |
15, 7388 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol,ethyl ether and chloroform | |
C1=CC=C(C=C1)C(=O)OC2=CC=CC=C2 | |
198.22 | |
CHEBI:86919 | |
99% | |
Phenyl benzoate, 99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-293-2
missing translation for 'rtecsNumber' : DH6299500
missing translation for 'tsca' : TSCA
RUO – Research Use Only