missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl carbamate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Phenyl carbamate, is used as various phenyl carbamate esters derived from amino acids, a dipeptide and amino acid esters and amides were prepared and assessed as potential prodrugs with the aim of protecting phenolic drugs against first-pass metabolism following per oral administration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenyl carbamate, is used as various phenyl carbamate esters derived from amino acids, a dipeptide and amino acid esters and amides were prepared and assessed as potential prodrugs with the aim of protecting phenolic drugs against first-pass metabolism following per oral administration.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Phenyl carbamate, is used as various phenyl carbamate esters derived from amino acids, a dipeptide and amino acid esters and amides were prepared and assessed as potential prodrugs with the aim of protecting phenolic drugs against first-pass metabolism following per oral administration.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 149°C to 152°C |
Quantity | 5 g |
Beilstein | 2042627 |
Solubility Information | Soluble in water. |
Formula Weight | 137.14 |
Percent Purity | ≥98% |
Chemical Name or Material | Phenyl carbamate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'