missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl chloroformate, 99%, Thermo Scientific Chemicals
£65.90 - £249.00
Chemical Identifiers
CAS | 1885-14-9 |
---|---|
Molecular Formula | C7H5ClO2 |
Molecular Weight (g/mol) | 156.57 |
MDL Number | MFCD00000637 |
InChI Key | AHWALFGBDFAJAI-UHFFFAOYSA-N |
Synonym | phenyl chloroformate, chloroformic acid phenyl ester, phenyl chlorocarbonate, carbonochloridic acid, phenyl ester, phenoxycarbonyl chloride, formic acid, chloro-, phenyl ester, fenylester kyseliny chlormravenci, phenylchloroformate, unii-6tnd0d6d3y, 6tnd0d6d3y |
PubChem CID | 15891 |
IUPAC Name | phenyl carbonochloridate |
SMILES | ClC(=O)OC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10124560
|
Thermo Scientific Acros
218212500 |
250 mL | Glass bottle |
£65.90
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11924941
|
Thermo Scientific Acros
218210010 |
1 L | Glass bottle |
£249.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentChemical Identifiers
1885-14-9 | |
156.57 | |
AHWALFGBDFAJAI-UHFFFAOYSA-N | |
15891 | |
ClC(=O)OC1=CC=CC=C1 |
C7H5ClO2 | |
MFCD00000637 | |
phenyl chloroformate, chloroformic acid phenyl ester, phenyl chlorocarbonate, carbonochloridic acid, phenyl ester, phenoxycarbonyl chloride, formic acid, chloro-, phenyl ester, fenylester kyseliny chlormravenci, phenylchloroformate, unii-6tnd0d6d3y, 6tnd0d6d3y | |
phenyl carbonochloridate |
Specifications
1885-14-9 | |
1.2440g/mL | |
80°C | |
98.5% min. (GC) | |
ClCO2C6H5 | |
MFCD00000637 | |
phenyl chloroformate, chloroformic acid phenyl ester, phenyl chlorocarbonate, carbonochloridic acid, phenyl ester, phenoxycarbonyl chloride, formic acid, chloro-, phenyl ester, fenylester kyseliny chlormravenci, phenylchloroformate, unii-6tnd0d6d3y, 6tnd0d6d3y | |
AHWALFGBDFAJAI-UHFFFAOYSA-N | |
phenyl carbonochloridate | |
15891 | |
156.57 | |
Liquid |
-28.0°C | |
188.0°C to 189.0°C (1013.0 mbar) | |
Authentic | |
C7H5ClO2 | |
1.5100 to 1.5120 | |
1.244 | |
Solubility in water: hydrolysis | |
ClC(=O)OC1=CC=CC=C1 | |
156.57 | |
1.86 mPa.s (20°C) | |
99% | |
Phenyl chloroformate |
Safety and Handling
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 217-547-8
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Phenyl chloroformate
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Acute toxicity Category 2
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Substances and mixtures corrosive to metals Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H330-Fatal if inhaled.
- H290-May be corrosive to metals.
- H412-Harmful to aquatic life with long lasting effects.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P402+P404-Store in a dry place. Store in a closed container.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title