missing translation for 'onlineSavingsMsg'
Learn More

Phenyl formate, 95%

Product Code. 11369973
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11369973

Brand: Thermo Scientific Alfa Aesar L00289.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1864-94-4 | C7H6O2 | 122.123 g/mol

Phenyl formate is widely used in the palladium-catalyzed carbonylation of aryl, alkenyl and allyl halides. It is also used as a reagent for the formylation of amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl formate is widely used in the palladium-catalyzed carbonylation of aryl, alkenyl and allyl halides. It is also used as a reagent for the formylation of amines.

Solubility
Immiscible with water, alcohol, ether and benzene.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1864-94-4
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.123
MDL Number MFCD00014126
InChI Key GEOWCLRLLWTHDN-UHFFFAOYSA-N
Synonym formic acid phenyl ester, formic acid, phenyl ester, phocho, formic acid, phenylester, acmc-1box2, phenyl formate gc
PubChem CID 74626
IUPAC Name phenyl formate
SMILES C1=CC=C(C=C1)OC=O

Specifications

Density 1.09
Boiling Point 72°C to 74°C (13 mmHg)
Flash Point 71°C (159°F)
Refractive Index 1.512
Quantity 5 g
Beilstein 507113
Solubility Information Immiscible with water,alcohol,ether and benzene.
Formula Weight 122.12
Percent Purity 95%
Chemical Name or Material Phenyl formate
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.