missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl formate, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Phenyl formate is widely used in the palladium-catalyzed carbonylation of aryl, alkenyl and allyl halides. It is also used as a reagent for the formylation of amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenyl formate is widely used in the palladium-catalyzed carbonylation of aryl, alkenyl and allyl halides. It is also used as a reagent for the formylation of amines.
Solubility
Immiscible with water, alcohol, ether and benzene.
Phenyl formate is widely used in the palladium-catalyzed carbonylation of aryl, alkenyl and allyl halides. It is also used as a reagent for the formylation of amines.
Solubility
Immiscible with water, alcohol, ether and benzene.
Specifications
Specifications
Density | 1.09 |
Boiling Point | 72°C to 74°C (13 mmHg) |
Flash Point | 71°C (159°F) |
Refractive Index | 1.512 |
Quantity | 25 g |
Beilstein | 507113 |
Solubility Information | Immiscible with water,alcohol,ether and benzene. |
Formula Weight | 122.12 |
Percent Purity | 95% |
Chemical Name or Material | Phenyl formate |
Safety and Handling
Product Identifier
- Phenyl formate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Phenyl formate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction