missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl phosphate, disodium salt dihydrate, 98%, Thermo Scientific Chemicals
Phenyl phosphate, disodium salt dihydrate, 98%, C6H5Na2O4P.2H2O, CAS Number-66778-08-3, sodiumphenylphosphatedibasicdihydrate, 10g, White, 221-917-4, 254.09, HYGROSCOPIC, UMRLIPYPVZYYFT-UHFFFAOYSA-N, Authentic, phenyl dihydrogen phosphate;sodium;dihydrate, C6H5OP(O)(ONa)2.2H2O, >300.0 deg.C
£20.40 - £103.00
Chemical Identifiers
CAS | 66778-08-3 |
---|---|
Molecular Formula | C6H5Na2O4P·2H2O |
Molecular Weight (g/mol) | 254.09 |
MDL Number | MFCD00149080 |
InChI Key | UMRLIPYPVZYYFT-UHFFFAOYSA-N |
Synonym | sodiumphenylphosphatedibasicdihydrate |
PubChem CID | 131883070 |
IUPAC Name | phenyl dihydrogen phosphate;sodium;dihydrate |
SMILES | C1=CC=C(C=C1)OP(=O)(O)O.O.O.[Na].[Na] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10184690
|
Thermo Scientific Acros
211830100 |
10 g | Glass bottle |
£20.40
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10318350
|
Thermo Scientific Acros
211831000 |
100 g | Glass bottle |
£103.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
66778-08-3 | |
254.09 | |
UMRLIPYPVZYYFT-UHFFFAOYSA-N | |
131883070 | |
C1=CC=C(C=C1)OP(=O)(O)O.O.O.[Na].[Na] |
C6H5Na2O4P·2H2O | |
MFCD00149080 | |
sodiumphenylphosphatedibasicdihydrate | |
phenyl dihydrogen phosphate;sodium;dihydrate |
Specifications
66778-08-3 | |
White | |
98% | |
C6H5OP(O)(ONa)2·2H2O | |
14, 3357 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol and chloroform,insoluble in acetone and ether | |
C1=CC=C(C=C1)OP(=O)(O)O.O.O.[Na].[Na] | |
254.09 | |
254.09 | |
Solid |
>300.0°C | |
Authentic | |
C6H5Na2O4P·2H2O | |
MFCD00149080 | |
sodiumphenylphosphatedibasicdihydrate | |
UMRLIPYPVZYYFT-UHFFFAOYSA-N | |
phenyl dihydrogen phosphate;sodium;dihydrate | |
131883070 | |
98% | |
Phenyl phosphate, disodium salt dihydrate, 98% |
Safety and Handling
HYGROSCOPIC
missing translation for 'einecsNumber' : 221-917-4
missing translation for 'tsca' : TSCA
RUO – Research Use Only