missing translation for 'onlineSavingsMsg'
Learn More

Phenyl salicylate, 99%, Thermo Scientific Chemicals

Product Code. 11419157
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Phenyl salicylate is used in the production of polymers, lacquers, adhesives, waxes and polishes. It acts as an antiseptic and mild analgesic. It is also used in the prevention of sunburn. It reacts with o-toluidine in 1,2,4-trichlorobenzene to give the corresponding amide o-Salicylotoluide. Further, it is involved in the study to check the effect on crystal size with cooling rate in the laboratory. In addition to this, it is used in veterinary medicine and also employed for coating pills, in which the medicine is intended for enteric release salol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl salicylate is used in the production of polymers, lacquers, adhesives, waxes and polishes. It acts as an antiseptic and mild analgesic. It is also used in the prevention of sunburn. It reacts with o-toluidine in 1,2,4-trichlorobenzene to give the corresponding amide o-Salicylotoluide. Further, it is involved in the study to check the effect on crystal size with cooling rate in the laboratory. In addition to this, it is used in veterinary medicine and also employed for coating pills, in which the medicine is intended for enteric release salol.

Solubility
Soluble in alcohol, ether, chloroform, turpentine, acetone and benzene. Sparingly soluble in chloroformbenzene. Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidants, strong acids and strong bases. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-55-8
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002213
InChI Key ZQBAKBUEJOMQEX-UHFFFAOYSA-N
Synonym phenyl salicylate, salol, phenol salicylate, salphenyl, musol, 2-phenoxycarbonylphenol, benzoic acid, 2-hydroxy-, phenyl ester, salicylic acid, phenyl ester, 2-hydroxybenzoic acid phenyl ester, phenyl-2-hydroxybenzoate
PubChem CID 8361
ChEBI CHEBI:34918
IUPAC Name phenyl 2-hydroxybenzoate
SMILES C1=CC=C(C=C1)OC(=O)C2=CC=CC=C2O

Specifications

Melting Point 41°C to 45°C
Boiling Point 172°C to 174°C (12 mmHg)
Flash Point 160°C (320°F)
Quantity 500 g
Beilstein 393969
Merck Index 14,7310
Solubility Information Soluble in alcohol,ether,chloroform,turpentine,acetone and benzene. Sparingly soluble in chloroformbenzene. Insoluble in water.
Formula Weight 214.22
Percent Purity 99%
Chemical Name or Material Phenyl salicylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'