missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetaldehyde, 95%

Product Code. p-7021786
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11404040

Brand: Thermo Scientific Alfa Aesar A14263.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 122-78-1 | C8H8O | 120.15 g/mol

Phenylacetaldehyde is used for the preparation of fragrances and polymers like polyesters, which find application as a rate controlling additive in polymerization reactions. It is an active component of fragrances and floral scent due to its honey-like sweet character and finds application in flavored cigarettes and beverages. It is an insect attractant and utilized in blacklight trap for pests. It is also used as a building block in the synthesis of more complex chemicals, such as resmethrin. Furthermore, it is used in association with acetic anhydride and allyltrimethylsilane in three-component coupling process catalyzed by LiBF4 providing homoallylic acetates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylacetaldehyde is used for the preparation of fragrances and polymers like polyesters, which find application as a rate controlling additive in polymerization reactions. It is an active component of fragrances and floral scent due to its honey-like sweet character and finds application in flavored cigarettes and beverages. It is an insect attractant and utilized in blacklight trap for pests. It is also used as a building block in the synthesis of more complex chemicals, such as resmethrin. Furthermore, it is used in association with acetic anhydride and allyltrimethylsilane in three-component coupling process catalyzed by LiBF4 providing homoallylic acetates.

Solubility
Miscible with alcohol and diethyl ether. Slightly miscible with water.

Notes
Store in cool place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-78-1
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.15
MDL Number MFCD00006993
InChI Key DTUQWGWMVIHBKE-UHFFFAOYSA-N
Synonym phenylacetaldehyde, benzeneacetaldehyde, hyacinthin, alpha-tolualdehyde, phenylethanal, 2-phenylethanal, phenylacetic aldehyde, alpha-toluic aldehyde, acetaldehyde, phenyl, benzylcarboxaldehyde
PubChem CID 998
ChEBI CHEBI:16424
IUPAC Name 2-phenylacetaldehyde
SMILES O=CCC1=CC=CC=C1

Specifications

Density 1.075
Boiling Point 194°C to 196°C
Flash Point 68°C (154°F)
Refractive Index 1.535
Quantity 25 g
Beilstein 385791
Sensitivity Air sensitive
Merck Index 14,7265
Solubility Information Miscible with alcohol and diethyl ether. Slightly miscible with water.
Formula Weight 120.15
Percent Purity 95%
Chemical Name or Material Phenylacetaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • Phenylacetaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Benzeneacetaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.