Learn More
Phenylacetaldehyde, 98%, stabilized, Thermo Scientific Chemicals
£34.10 - £108.00
Chemical Identifiers
CAS | 122-78-1 |
---|---|
Molecular Formula | C8H8O |
Molecular Weight (g/mol) | 120.15 |
MDL Number | MFCD00006993 |
InChI Key | DTUQWGWMVIHBKE-UHFFFAOYSA-N |
Synonym | phenylacetaldehyde, benzeneacetaldehyde, hyacinthin, alpha-tolualdehyde, phenylethanal, 2-phenylethanal, phenylacetic aldehyde, alpha-toluic aldehyde, acetaldehyde, phenyl, benzylcarboxaldehyde |
PubChem CID | 998 |
ChEBI | CHEBI:16424 |
IUPAC Name | 2-phenylacetaldehyde |
SMILES | O=CCC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10058311
|
Thermo Scientific
370911000 |
100 g | Glass bottle |
£34.10
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10292332
|
Thermo Scientific Acros
370915000 |
500 g | Glass bottle |
£108.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
122-78-1 | |
120.15 | |
DTUQWGWMVIHBKE-UHFFFAOYSA-N | |
998 | |
2-phenylacetaldehyde |
C8H8O | |
MFCD00006993 | |
phenylacetaldehyde, benzeneacetaldehyde, hyacinthin, alpha-tolualdehyde, phenylethanal, 2-phenylethanal, phenylacetic aldehyde, alpha-toluic aldehyde, acetaldehyde, phenyl, benzylcarboxaldehyde | |
CHEBI:16424 | |
O=CCC1=CC=CC=C1 |
Specifications
5mg KOH/g max. | |
Colorless to Yellow | |
193.0°C | |
Authentic | |
C8H8O | |
1.5250 to 1.5320 | |
07, 292 | |
1.029 | |
Solubility in water: 2.210g/L water (25°C). Other solubilities: soluble in ethanol,acetone,diethyl ether | |
O=CCC1=CC=CC=C1 | |
approx. 0.01% citric acid | |
998 | |
120.15 | |
Liquid |
122-78-1 | |
1.0290g/mL | |
68°C | |
97.5% min. (GC) | |
C6H5CH2CHO | |
MFCD00006993 | |
15, 7377 | |
phenylacetaldehyde, benzeneacetaldehyde, hyacinthin, alpha-tolualdehyde, phenylethanal, 2-phenylethanal, phenylacetic aldehyde, alpha-toluic aldehyde, acetaldehyde, phenyl, benzylcarboxaldehyde | |
DTUQWGWMVIHBKE-UHFFFAOYSA-N | |
2-phenylacetaldehyde | |
120.15 | |
CHEBI:16424 | |
98% | |
Phenylacetaldehyde, 98% |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If s
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 204-574-5
Recommended Storage : Refrigerator +4°C
- Phenylacetaldehyde, stabilized
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
RUO – Research Use Only