missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylacetylene, 98%, pure
Tous les produits Thermo Scientific Chemicals Produits
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Conditionnement:
100g
25g
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Melting Point | -44.8°C |
| Color | Colorless to Yellow |
| Density | 0.9300g/mL |
| Boiling Point | 142.0°C to 144.0°C |
| Flash Point | 31°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | C6H5C≡CH |
| Refractive Index | 1.5470 to 1.5500 |
| Afficher plus |
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu