missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylazoformic acid 2-phenylhydrazide compound with 1,5-Diphenylcarbohydrazide, ACS, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Phenylazoformic acid 2-phenylhydrazide compound with 1,5-Diphenylcarbohydrazide is used as an indicator in titrating iron and for the colorimetric determination of chromium and the detection of cadmium, mercury, magnesium, aldehydes, and emetine, a color developing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 154°C to 156°C |
Color | Orange |
Odor | Odorless |
Quantity | 50 g |
Beilstein | 6110063 |
Merck Index | 14,3322 |
Formula Weight | 482.55 |
Grade | ACS |
Physical Form | Powder |
Chemical Name or Material | Phenylazoformic acid 2-phenylhydrazide compound, with 1,5-Diphenylcarbohydrazide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'