missing translation for 'onlineSavingsMsg'
Learn More
Phenylmagnesium bromide, 1.6M solution in CPME, AcroSeal™, Thermo Scientific Chemicals Product Code.: 12635017

Phenylmagnesium bromide, 1.6M solution in CPME, AcroSeal™, Thermo Scientific Chemicals

Product Code. 12635017
800 mL, AcroSeal™ Glass bottle
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy

Product Code. 12635017

Brand: Thermo Scientific Acros 445968000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-58-3, 5614-37-9
Molecular Formula C6H5BrMg
Molecular Weight (g/mol) 181.32
MDL Number MFCD00000038
InChI Key ANRQGKOBLBYXFM-UHFFFAOYSA-M
Synonym phenylmagnesium bromide, bromophenylmagnesium, magnesium, bromophenyl, phmgbr, phenylmagnesiumbromide, bromo phenyl magnesium, phenyl magnesium bromide, phenylmagnesium bromide solution, 3.0 m in diethyl ether, bromomagnesiobenzene, phenyhnagnesium bromide, grignard reagentShow MoreShow Less
PubChem CID 66852
IUPAC Name magnesium;benzene;bromide
SMILES Br[Mg]C1=CC=CC=C1

Specifications

Flash Point −1°C
Packaging AcroSeal™ Glass bottle
Linear Formula C6H5MgBr
Quantity 800 mL
Solubility Information Solubility in water: reacts
Formula Weight 181.31
Chemical Name or Material Phenylmagnesium bromide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0453760 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number445961000,445968000
Lot NumberA0403002 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number445961000,445968000
Lot NumberA0429739 Certificate TypeCertificate of Analysis Date12/05/2021 Catalog Number
Lot NumberA0412227 Certificate TypeCertificate of Analysis Date07/10/2020 Catalog Number445961000,445968000
Lot NumberA0412019 Certificate TypeCertificate of Analysis Date29/09/2020 Catalog Number445961000,445968000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenylmagnesium bromide, 1.6M solution in CPME
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phenylmagnesium bromide, 1.6M solution in CPME, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.