Learn More
Thermo Scientific Chemicals Phenylmethanesulfonyl fluoride, 99%
£30.90 - £293.00
Chemical Identifiers
CAS | 329-98-6 |
---|---|
Molecular Formula | C7H7FO2S |
Molecular Weight (g/mol) | 174.19 |
MDL Number | MFCD00007424 |
InChI Key | YBYRMVIVWMBXKQ-UHFFFAOYSA-N |
Synonym | phenylmethylsulfonyl fluoride, pmsf, benzenemethanesulfonyl fluoride, benzylsulfonyl fluoride, alpha-toluenesulfonyl fluoride, benzylsulphonyl fluoride, alpha-toluenesulphonyl fluoride, phenylmethylsulfonylfluoride, phenylmethylsulphonyl fluoride, .alpha.-toluenesulfonyl fluoride |
PubChem CID | 4784 |
ChEBI | CHEBI:8102 |
IUPAC Name | phenylmethanesulfonyl fluoride |
SMILES | C1=CC=C(C=C1)CS(=O)(=O)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10358110
|
Thermo Scientific Chemicals
215740010 |
1 g | Glass bottle |
£30.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10042680
|
Thermo Scientific Chemicals
215740050 |
5 g | Glass bottle |
£46.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10105180
|
Thermo Scientific Chemicals
215740100 |
10 g | Glass bottle |
£76.60
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10267023
|
Thermo Scientific Chemicals
215740500 |
50 g | Glass bottle |
£293.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
329-98-6 | |
174.19 | |
YBYRMVIVWMBXKQ-UHFFFAOYSA-N | |
4784 | |
phenylmethanesulfonyl fluoride |
C7H7FO2S | |
MFCD00007424 | |
phenylmethylsulfonyl fluoride, pmsf, benzenemethanesulfonyl fluoride, benzylsulfonyl fluoride, alpha-toluenesulfonyl fluoride, benzylsulphonyl fluoride, alpha-toluenesulphonyl fluoride, phenylmethylsulfonylfluoride, phenylmethylsulphonyl fluoride, .alpha.-toluenesulfonyl fluoride | |
CHEBI:8102 | |
C1=CC=C(C=C1)CS(=O)(=O)F |
Specifications
329-98-6 | |
100.0 | |
White | |
98.5% min. (GC) | |
C6H5CH2SO2F | |
11, III, 331 | |
phenylmethylsulfonyl fluoride, pmsf, benzenemethanesulfonyl fluoride, benzylsulfonyl fluoride, alpha-toluenesulfonyl fluoride, benzylsulphonyl fluoride, alpha-toluenesulphonyl fluoride, phenylmethylsulfonylfluoride, phenylmethylsulphonyl fluoride, .alpha.-toluenesulfonyl fluoride | |
YBYRMVIVWMBXKQ-UHFFFAOYSA-N | |
phenylmethanesulfonyl fluoride | |
4784 | |
174.19 | |
Needles (May Agglomerate) |
98.5 | |
90.0°C to 94.0°C | |
Authentic | |
C7H7FO2S | |
MFCD00007424 | |
15, 7659 | |
Solubility in water: hydrolysis. Other solubilities: soluble in xylene and petroleum ether | |
C1=CC=C(C=C1)CS(=O)(=O)F | |
174.19 | |
CHEBI:8102 | |
99% | |
Phenylmethanesulfonyl fluoride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minut
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 206-350-2
- Phenylmethanesulfonyl fluoride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only