Learn More
Phenylphosphonic acid, 98%, Thermo Scientific Chemicals
£32.70 - £361.00
Chemical Identifiers
CAS | 1571-33-1 |
---|---|
Molecular Formula | C6H7O3P |
Molecular Weight (g/mol) | 158.09 |
MDL Number | MFCD00002136 |
InChI Key | QLZHNIAADXEJJP-UHFFFAOYSA-N |
Synonym | benzenephosphonic acid, phosphonic acid, phenyl, phenyl-phosphonic acid, phosphonic acid, p-phenyl, phenyl phosphonic acid, unii-byd76t2868, sv7, pubchem21303, phosphonic acid,phenyl, wln: qpqo&r |
PubChem CID | 15295 |
IUPAC Name | phenylphosphonic acid |
SMILES | OP(O)(=O)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10502151
|
Thermo Scientific Acros
130791000 |
100 g | Glass bottle |
£32.70
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10497710
|
Thermo Scientific Acros
130795000 |
500 g | Glass bottle |
£107.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10534074
|
Thermo Scientific Acros
130790025 |
2.5 kg | Plastic Bottle |
£361.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1571-33-1 | |
158.09 | |
QLZHNIAADXEJJP-UHFFFAOYSA-N | |
15295 | |
OP(O)(=O)C1=CC=CC=C1 |
C6H7O3P | |
MFCD00002136 | |
benzenephosphonic acid, phosphonic acid, phenyl, phenyl-phosphonic acid, phosphonic acid, p-phenyl, phenyl phosphonic acid, unii-byd76t2868, sv7, pubchem21303, phosphonic acid,phenyl, wln: qpqo&r | |
phenylphosphonic acid |
Specifications
1571-33-1 | |
White | |
98% | |
C6H5P(O)(OH)2 | |
16,803 | |
Solubility in water: 40.4g/100mL water (25°C). Other solubilities: soluble in alcohol and ccl4 | |
OP(O)(=O)C1=CC=CC=C1 | |
158.09 | |
158.09 | |
Crystalline Powder |
158.0°C to 165.0°C | |
Authentic | |
C6H7O3P | |
MFCD00002136 | |
benzenephosphonic acid, phosphonic acid, phenyl, phenyl-phosphonic acid, phosphonic acid, p-phenyl, phenyl phosphonic acid, unii-byd76t2868, sv7, pubchem21303, phosphonic acid,phenyl, wln: qpqo&r | |
QLZHNIAADXEJJP-UHFFFAOYSA-N | |
phenylphosphonic acid | |
15295 | |
98% | |
Phenylphosphonic acid, 98% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 216-388-1
missing translation for 'rtecsNumber' : TA0350000
missing translation for 'tsca' : TSCA
- Phenylphosphonic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only