Learn More
Phenylsilane, 97%, Thermo Scientific Chemicals
£58.89 - £743.70
Chemical Identifiers
CAS | 694-53-1 |
---|---|
Molecular Formula | C6 H8 Si |
Molecular Weight (g/mol) | 108.22 |
MDL Number | MFCD00013478 |
InChI Key | XJWOWXZSFTXJEX-UHFFFAOYSA-N |
Synonym | phenylsilane, benzene, silyl, silylbenzene, silane, phenyl, fenylsilan, fenylsilan czech, phenylsilane, silyl, silane, phenyl-, |
PubChem CID | 6327628 |
IUPAC Name | phenylsilicon |
SMILES | C1=CC=C(C=C1)[Si] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11499006
|
Thermo Scientific Alfa Aesar
A17901.06 |
5 g |
£58.89
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409016
|
Thermo Scientific Alfa Aesar
A17901.14 |
25 g |
£197.99
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419016
|
Thermo Scientific Alfa Aesar
A17901.22 |
100 g |
£743.70
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical intermediate.
Solubility
Contact with water releases a flammable alcohol. Soluble in water (Contact with water releases flammable gases), and most common organic solvents.
Notes
Moisture Sensitive. Store away from water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect against electrostatic charges. Keep ignition sources away. Store under inert gas.
Chemical Identifiers
694-53-1 | |
108.22 | |
XJWOWXZSFTXJEX-UHFFFAOYSA-N | |
6327628 | |
C1=CC=C(C=C1)[Si] |
C6 H8 Si | |
MFCD00013478 | |
phenylsilane, benzene, silyl, silylbenzene, silane, phenyl, fenylsilan, fenylsilan czech, phenylsilane, silyl, silane, phenyl-, | |
phenylsilicon |
Specifications
120°C | |
C6 H8 Si | |
UN3399 | |
phenylsilane, benzene, silyl, silylbenzene, silane, phenyl, fenylsilan, fenylsilan czech, phenylsilane, silyl, silane, phenyl-, | |
XJWOWXZSFTXJEX-UHFFFAOYSA-N | |
phenylsilicon | |
6327628 | |
97% | |
Moisture sensitive | |
Phenylsilane |
694-53-1 | |
MFCD00013478 | |
7°C (45°F) | |
Contact with water releases a flammable alcohol. Soluble in water (Contact with water releases flammable gases),and most common organic solvents. | |
C1=CC=C(C=C1)[Si] | |
108.22 | |
108.22 | |
1.5105 | |
0.88 |
Safety and Handling
GHS H Statement
H225-H261-H302-H332-H315-H319-H335
Highly flammable liquid and vapour.
In contact with water releases flammable gas.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P335+P334-P363-P370+P378q-P50
H225-H261-H302+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
missing translation for 'einecsNumber' : 211-772-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Phenylsilane
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H261-In contact with water releases flammable gases.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- EUH014-Reacts violently with water.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Phenylsilane
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.