missing translation for 'onlineSavingsMsg'
Learn More
Phenyltin trichloride, Thermo Scientific Chemicals Product Code.: 11364849

Phenyltin trichloride, Thermo Scientific Chemicals

Product Code. 11364849
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11364849

Brand: Thermo Scientific Alfa Aesar 071157.14

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phenyltin trichloride is used to produce 3-phenyl-cyclohexanone with cyclohex-2-enone. And it is also used in pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1124-19-2
Molecular Formula C6H5Cl3Sn
Molecular Weight (g/mol) 302.166
MDL Number MFCD00000513
InChI Key UBOGEXSQACVGEC-UHFFFAOYSA-K
Synonym phenyltin trichloride, phenyltrichlorotin, stannane, trichlorophenyl, trichlorophenyltin, trichloro phenyl stannane, phenyltrichlorostannane, ccris 6323, phsncl3, phenyl tin trichloride, monophenyltin trichloride
PubChem CID 14298
IUPAC Name trichloro(phenyl)stannane
SMILES C1=CC=C(C=C1)[Sn](Cl)(Cl)Cl

Specifications

Density 1.84 g/mL
Odor Pungent
Quantity 25 g
UN Number UN2788
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 302.18
Physical Form Liquid
Chemical Name or Material Phenyltin trichloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ20J013 Certificate TypeCertificate of Analysis Date16/01/2024 Catalog Number071157.14,071157.06
Lot NumberL28U055 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number071157.14,071157.06
Lot NumberQ12E017 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number071157.14,071157.06
Lot NumberZ27E020 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number071157.14,071157.06
Lot NumberJ07J39 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number071157.14,071157.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phenyltin trichloride, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.