missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrimethylammonium hydroxide, 0.1 M in methanol

Product Code. p-184247 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10781832

Brand: Thermo Scientific Acros 422021000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1899-02-1 | C9H15NO | 153.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenyltrimethylammonium hydroxide
Color Colorless
Name Note 0.1M in Methanol
CAS 1899-02-1, 67-56-1
Molecular Formula C9H15NO
Linear Formula C6H5N(CH3)3OH
MDL Number MFCD00041899
Concentration 0.09 to 0.11M
Density 0.8000g/mL
Synonym methelute, phenyltrimethylammonium hydroxide, trimethylanilinium hydroxide, phenyl trimethyl ammonium hydroxide, trimethylphenylammonium hydroxide, n,n,n-trimethylbenzenaminium hydroxide, n,n,n-trimethylanilinium hydroxide, benzenaminium, n,n,n-trimethyl-, hydroxide, ptah, ammonium, phenyltrimethyl-, hydroxide
InChI Key HADKRTWCOYPCPH-UHFFFAOYSA-M
SMILES [OH-].C[N+](C)(C)C1=CC=CC=C1
IUPAC Name N,N,N-trimethylanilinium hydroxide
Molecular Weight (g/mol) 153.23
PubChem CID 15913
ChEBI CHEBI:85062
Formula Weight 153.22
Packaging Glass bottle
Specific Gravity 0.8
Quantity 100 g
Flash Point 10°C
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenyltrimethylammonium hydroxide, 0.1 M in methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H370-Causes damage to organs.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.