Learn More
Uvex™ Pheos B-WR Safety Helmet
Helmets are resistant to hot molten metal splash and feature a comfortable, secure fit with continuous width adjustment.
£0.59 - £13.77
Specifications
Model Pheos B-WR
Ventilated Ventilated
View More Specs
Products 2
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
15366222
View Documents Promotion Details
Uvex™
9772930
Black This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15306212
View Documents Promotion Details
Uvex™
9772030
White This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Resistant to hot molten metal splash and temperatures down to -30°C (EN 367 additional requirement MM)
  • Side euroslot adapters (30 mm) are suitable for the addition of earmuffs or the uvex pheos visor system
  • Suspension harness with wheel ratchet allows continuous width adjustment, for a perfect fit at all times
  • Six-point textile band attachment ensures optimal fit and comfort
  • Available in a wide variety of color options
Specifications
Specifications
Pheos B-WR
Ventilated