Learn More
Thermo Scientific™ phi29 DNA Polymerase (10 U/μL)
Thermo Scientific phi29 DNA Polymerase is a highly processive polymerase featuring strong strand displacement activity, which allows for highly efficient isothermal DNA amplification.
£82.25 - £1064.00
Specifications
Product Type | Stand-alone enzyme |
---|---|
For Use With (Application) | MDA-WGA, RCA |
Polymerase | phi29 DNA Polymerase |
Optimal Reaction Temperature | 30°C |
Shipping Condition | Dry Ice |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10233130
|
Thermo Scientific™
EP0091 |
250 units |
£82.25
250 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10242410
|
Thermo Scientific™
EP0092 |
1,000 units |
£270.00
1000 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10775271
|
Thermo Scientific™
EP0094 |
5,000 units |
£1064.00
5000 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thermo Scientific phi29 DNA Polymerase is a highly processive polymerase featuring strong strand displacement activity, which allows for highly efficient isothermal DNA amplification. Produced from E. coli cells with a cloned gene 2 of Bacillus subtilis phage phi29, phi29 DNA Polymerase possesses 3'→5' proofreading exonuclease activity that acts preferentially on single-stranded DNA or RNA.
Thermo Scientific phi29 DNA Polymerase is a highly processive polymerase featuring strong strand displacement activity, which allows for highly efficient isothermal DNA amplification. Produced from E. coli cells with a cloned gene 2 of Bacillus subtilis phage phi29, phi29 DNA Polymerase possesses 3'→5' proofreading exonuclease activity that acts preferentially on single-stranded DNA or RNA.
Features of phi29 DNA Polymerase
• Excellent processivity and strand displacement activity—more than 70 kb long DNA stretches can be synthesized
• Highly accurate DNA synthesis
• High yields of amplified DNA even from small amounts of template
Applications
Phi29 DNA Polymerase may be used in a wide variety of applications including:
• Rolling circle amplification (RCA)—generation of periodic DNA nanotemplates
• Multiple displacement amplification (MDA)
• Unbiased amplification of whole genome (WGA) (see figure below)
• Amplification of DNA for SNP and STR detection
• Cell-free amplification of DNA from single cells, pathogenic organisms, or metagenomes
• DNA template preparation for sequencing
• Protein-primed DNA amplification
• Recombination based-cloning
• Cell-free cloning of lethal DNA
• RNA-primed DNA amplification
Notes
1. The addition of pyrophosphatase to the reaction mixture with phi29 DNA Polymerase may enhance DNA synthesis.
2. Because of the enzymes’s 3’→5’ proofreading exonuclease, 3'-modified primers are highly recommended.
Specifications
Stand-alone enzyme | |
phi29 DNA Polymerase | |
Dry Ice | |
4 hours |
MDA-WGA, RCA | |
30°C | |
10 U/μL |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.