Learn More
Thermo Scientific™ Phire Animal Tissue Direct PCR Kit
Perform PCR directly from various animal tissues including mice, fish, birds and insects without DNA purification.
Brand: Thermo Scientific™ F-140
Code : 87
Additional Details : Weight : 0.10000kg
Description
Thermo Scientific Phire Animal Tissue Direct PCR Kit has been developed for amplification of DNA directly from a wide variety of animal tissues including mice, fish, birds and insects.The kit contains all the necessary components for amplification of DNA directly from animal tissues: optimized reagents, sampling tools, and DNARelease Additive, which can be used to improve the release of DNA from animal tissues. In addition, the kit includes detailed instructions and control primers that can be used with numerous animal species.
Highlights
- No need for time-consuming and expensive DNA purification steps
- Very little sample material required
- Two simple protocols for various applications
- Convenient sampling tools included in the kit
- Phire Hot Start II DNA Polymerase provides high yields of specific product with short extension time (20 s/kb).
Specifications
Phire Hot Start II DNA Polymerase, 2x Phire Animal Tissue PCR Buffer (includes dNTPs and MgCl2), Dilution Buffer, DNA Release Additive, Gel loading dye, Harris Uni-Core 0.50 mm, Harris Cutting Mat, Universal control primer mix. Recommended storage temperature -20°C | |
Phire™ Animal Tissue Direct PCR Kit | |
200 x 50µL Rxns | |
Genotyping |