Learn More
Thermo Scientific™ Phire Hot Start II DNA Polymerase
Thermo Scientific Phire Hot Start II DNA Polymerase is an enhanced PCR enzyme for routine and high throughput PCR applications.
£136.00 - £576.00
Specifications
GC-Rich PCR Performance | Medium |
---|---|
Polymerase | Phire Hot Start II DNA Polymerase |
Reaction Speed | Fast |
Product Type | DNA Polymerase |
Purity or Quality Grade | DHPLC |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10185667
|
Thermo Scientific™
F122S |
200 Reactions |
£136.00
200 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10247467
|
Thermo Scientific™
F122L |
1000 Reactions |
£576.00
1000 reactions |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Thermo Scientific Phire Hot Start II DNA Polymerase is an enhanced PCR enzyme for routine and high throughput PCR applications. It outperforms every Taq-based hot start polymerase on the market. Phire Hot Start II DNA Polymerase is significantly faster, extremely robust, and also capable of amplifying long DNA fragments with high yields. These features are achieved through advanced protein engineering of the polymerase. It incorporates a unique double-stranded DNA binding domain which allows short extension times (10 to 15 s/kb), improves yields, and increases fidelity 2-fold compared to Taq DNA polymerase (Cat. No. EP0072).
The hot start modification of Phire Hot Start II DNA Polymerase is based on the Affibody inactivation technology. This technology increases the specificity of the PCR reaction with no additional time required for initial activation of the enzyme.
Highlights
- Quick hot start—No reactivation step
- Fast enzyme—Amplify four times faster than with hot start Taq
- Robust—Minimal reaction optimization due to high inhibitor tolerance
- High yields—Abundant products due to high efficiency
- Longer PCR products—Amplify significantly longer DNA fragments than with any hot start Taq
Applications
- Hot Start PCR
- Routine PCR
- Non-high fidelity PCR
- Fast PCR
- High throughput PCR
- Genotyping
Note:The optimal annealing temperature for Phire DNA Polymerases may differ significantly from that of Taq-based polymerases. For optimal results start by accurately calculating your Tm with our Tm calculator.
Phire Green Hot Start II DNA Polymerase is also available (Cat. No. F-124S or F-124L) for direct loading of PCR products on gel.
Specifications
Medium | |
Fast | |
DHPLC | |
Cloning, Hot-start PCR | |
Built-In Hot Start | |
Separate Components |
Phire Hot Start II DNA Polymerase | |
DNA Polymerase | |
Wet or Dry Ice | |
2X | |
Blunt | |
DNA |
For Research Use Only. Not for use in diagnostic procedures.