missing translation for 'onlineSavingsMsg'
Learn More

Phloroglucide, 95%, Thermo Scientific Chemicals

Product Code. 11455337
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 491-45-2
Molecular Formula C12H10O5
Molecular Weight (g/mol) 234.21
MDL Number MFCD01543374
InChI Key KICYRZIVKKYRFS-UHFFFAOYSA-N
Synonym phloroglucide, 2,3',4,5',6-biphenylpentol, 2-3,5-dihydroxyphenyl benzene-1,3,5-triol, 1,1'-biphenyl-2,3',4,5',6-pentol, phloroglucide hydrate, 1,1'-biphenyl-2,3',4,5',6-pentaol, phloroglucid, d0o8yz, 2,4,5',6-biphenylpentol
PubChem CID 248349
SMILES OC1=CC(O)=C(C(O)=C1)C1=CC(O)=CC(O)=C1

Specifications

Melting Point ∼296°C (decomposition)
Quantity 100 g
Beilstein 3324960
Sensitivity Light sensitive
IUPAC Name 2-(3,5-dihydroxyphenyl)benzene-1,3,5-triol
Formula Weight 234.21
Percent Purity 95%
Chemical Name or Material Phloroglucide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'