missing translation for 'onlineSavingsMsg'
Learn More
Phloroglucinol Dihydrate, Certified AR for Analysis, Fisher Chemical Product Code.: 10151770

Phloroglucinol Dihydrate, Certified AR for Analysis, Fisher Chemical

Product Code. 10151770
25 g, Plastic powder jar
Click to view available options
Quantity:
25 g
Packaging:
Plastic powder jar
This item is not returnable. View return policy

Product Code. 10151770

Brand: Fisher Chemical P/2922/46

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 6099-90-7
Melting Point 218°C
Color Cream
Packaging Plastic powder jar
Molecular Formula C6H10O5
MDL Number 149090
Quantity 25 g
Synonym phloroglucinol dihydrate, benzene-1,3,5-triol dihydrate, 1,3,5-trihydroxybenzene dihydrate, 1,3,5-benzenetriol, dihydrate, phloroglucinol, dihydrate, sym-trihydroxybenze, unii-0x7b22brg1, ccris 6521, 1,3,5-benzoltrioldihydrat, phloroglucinoldihydrate
InChI Key MPYXTIHPALVENR-UHFFFAOYSA-N
SMILES C1=C(C=C(C=C1O)O)O.O.O
IUPAC Name benzene-1,3,5-triol;dihydrate
Molecular Weight (g/mol) 162.141
PubChem CID 80196
Formula Weight 162.14g/mol
Physical Form Powder
Show More Show Less
compliance-icons
Product Identifier
  • Phloroglucinol dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phloroglucinol Dihydrate, Certified AR for Analysis, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.