missing translation for 'onlineSavingsMsg'
Learn More

Phorbol 12-myristate 13-acetate, 97%

Product Code. 10515491 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 mg
5 mg
Packaging:
Glass bottle
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 10515491

Brand: Thermo Scientific 356150010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16561-29-8 | C36H56O8 | 616.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16561-29-8
Molecular Formula C36H56O8
Molecular Weight (g/mol) 616.84
MDL Number MFCD00036736
InChI Key PHEDXBVPIONUQT-RGYGYFBISA-N
Synonym phorbol 12-myristate 13-acetate, phorbol ester, 12-o-tetradecanoylphorbol-13-acetate, 12-o-tetradecanoylphorbol 13-acetate, factor a1, tetradecanoylphorbol acetate, 12-tetradecanoylphorbol 13-acetate, factor a1 croton oil, phorbol 13-acetate 12-myristate, phorbol-12-myristate-13-acetate
PubChem CID 27924
ChEBI CHEBI:37537
IUPAC Name (1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O

Specifications

Melting Point 72.0°C
Color White to Yellow
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 mg
Merck Index 14,7332
Formula Weight 616.84
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material Phorbol 12-myristate 13-acetate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phorbol 12-myristate 13-acetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.