missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phorbol 12-myristate 13-acetate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 mg
5 mg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 72.0°C |
Color | White to Yellow |
Assay Percent Range | 96% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 5 mg |
Merck Index | 14,7332 |
Formula Weight | 616.84 |
Percent Purity | 97% |
Physical Form | Fine Crystalline Powder |
Chemical Name or Material | Phorbol 12-myristate 13-acetate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0468483 | Certificate of Analysis | 31/01/2025 | 356150010,356150050 |
A0467177 | Certificate of Analysis | 31/01/2025 | 356150010,356150050 |
A01815 | Certificate of Analysis | 07/11/2024 | 356150010,356150050 |
A0464427 | Certificate of Analysis | 19/07/2024 | 356150010,356150050 |
A0460868 | Certificate of Analysis | 11/06/2024 | 356150010,356150050 |
Safety and Handling



Product Identifier
- Phorbol 12-myristate 13-acetate
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H351-Suspected of causing cancer.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Phorbol 12-myristate 13-acetate, 97%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction