missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phorbol 12-myristate 13-acetate, 95%, Thermo Scientific Chemicals
64.98 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Description
PMA activates Ca2+- ATPase and potentiates forskolin-induced cAMP formation. It has been shown to inhibit apoptosis induced by the Fas antigen, but PMA induces apoptosis in HL-60 promyelocytic leukemia cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
PMA activates Ca2+- ATPase and potentiates forskolin-induced cAMP formation. It has been shown to inhibit apoptosis induced by the Fas antigen, but PMA induces apoptosis in HL-60 promyelocytic leukemia cells.
Solubility
Very soluble in methanol, soluble in DMSO, chloroform, ethanol or acetone, ether, ethyl acetate, DMF or acetonitrile. Insoluble in water.
Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
PMA activates Ca2+- ATPase and potentiates forskolin-induced cAMP formation. It has been shown to inhibit apoptosis induced by the Fas antigen, but PMA induces apoptosis in HL-60 promyelocytic leukemia cells.
Solubility
Very soluble in methanol, soluble in DMSO, chloroform, ethanol or acetone, ether, ethyl acetate, DMF or acetonitrile. Insoluble in water.
Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | White |
Quantity | 1 mg |
Beilstein | 2407201 |
Sensitivity | Light sensitive |
Merck Index | 14,7332 |
Solubility Information | Very soluble in methanol,soluble in DMSO,chloroform,ethanol or acetone,ether,ethyl acetate,DMF or acetonitrile. Insoluble in water. |
Formula Weight | 616.83 |
Percent Purity | ≥99% |
Physical Form | Film or Powder |
Chemical Name or Material | Phorbol 12-myristate 13-acetate |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Phorbol 12-myristate 13-acetate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Tetradecanoyl phorbol acetate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'