missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Phosphate, 0.2M buffer soln., pH 8.5
Used in molecular biology, biochemistry, and chromatography. It is used in the purification of antibodies.
£34.10 - £43.50
Specifications
Concentration | 0.2M buffer soln. |
---|---|
Molecular Weight (g/mol) | 119.976 |
Packaging | Plastic Bottle |
CAS | 7558-80-7 |
Content And Storage | Ambient Temperatures |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
0.2M buffer soln. | |
Plastic Bottle | |
Ambient Temperatures |
119.976 | |
7558-80-7 | |
Liquid |