Learn More
Thermo Scientific Chemicals Phosphate-buffered saline (DPBS, 1X), Dulbecco's formula, without calcium, without magnesium
£30.05 - £45.02
Specifications
Content And Storage | Ambient temperatures |
---|---|
Packaging Type | Plastic bottle |
Form | Liquid |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
15434695
|
Thermo Scientific Chemicals
J67802.AP |
500 mL |
£30.05
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15444695
|
Thermo Scientific Chemicals
J67802.K2 |
1 L |
£45.02
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
An anticoagulant and blood preservative. Permits the storage of whole blood at refrigerator temperatures for approximately 10 weeks.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ambient temperatures | |
Plastic bottle | |
Liquid |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.