Learn More
Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 10X), RNase free
Helps to maintain a constant pH
£81.60 - £131.00
Specifications
Dilution | 10 X |
---|---|
Product Type | Phosphate Buffered Saline (PBS) |
Form | Liquid |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
15490267
|
Thermo Scientific Chemicals
J62851.AP |
500 mL |
£131.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15480267
|
Thermo Scientific Chemicals
J62851.AK |
250 mL |
£81.60
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phosphate-buffered saline (PBS, 10X), is an isotonic, non-toxic to most cells and is used in biological research. Phosphate-buffered saline with EDTA is used to disengage attached and clumped cells. It is also used in cell culturing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
10 X | |
Phosphate Buffered Saline (PBS) | |
Liquid |
Safety and Handling
Recommended Storage : Ambient temperatures
RUO – Research Use Only